(4-Methoxycarbonylbenzyl)triphenylphosphoniumbromid

Lieferant: BIONET RESEARCH

Synonyme: [4-(Methoxycarbonyl)benzyl](triphenyl)phosphonium bromide

7K-902.500 7K-902.1 7K-902.5 7K-902.10
BIOT7K-902.500EA 0 EUR
BIOT7K-902.500 BIOT7K-902.1 BIOT7K-902.5 BIOT7K-902.10
(4-Methoxycarbonylbenzyl)triphenylphosphoniumbromid
(4-Methoxycarbonylbenzyl)triphenylphosphoniumbromid
Formel: C₂₇H₂₄BrO₂P
Molecular Weight: 491.37 g/mol
MDL: MFCD00082765
CAS-Nummer: 1253-46-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR