(4-Nitrobenzyl)triphenylphosphoniumchlorid ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(4-Nitrobenzyl)triphenylphosphonium bromide
A15288.06
A15288.14
A15288.06EA
23.8
EUR
A15288.06
A15288.14
(4-Nitrobenzyl)triphenylphosphoniumchlorid ≥98%
(4-Nitrobenzyl)triphenylphosphoniumchlorid
Purity: 98%
CAS No.: 2767-70-6
Molecular Formula: C25H21BrNO2P
Formula Weight: 478.33
Melting Point: ca 280° dec.
MDL No.: MFCD00032108
Formel:
C₂₅H₂₁BrNO₂P Molecular Weight: 478,33 g/mol Schmelzpunkt: ∼ 280 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00032108 CAS-Nummer: 2767-70-6 EINECS: 220-444-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...