(4-Nitrobenzyl)triphenylphosphoniumchlorid
Lieferant: CHEMPUR
Synonyme:
(4-Nitrobenzyl)triphenylphosphonium bromide
NRB00503.250
NRB00503.1
CHMPNRB00503.250EA
0
EUR
CHMPNRB00503.250
MAYBNRB00503.1
(4-Nitrobenzyl)triphenylphosphoniumchlorid
(4-Nitrobenzyl)triphenylphosphoniumchlorid
Formel:
C₂₅H₂₁BrNO₂P Molecular Weight: 478.33 g/mol |
MDL:
MFCD00032108 CAS-Nummer: 2767-70-6 EINECS: 220-444-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...