Alcianblau 8 GS

Lieferant: Serva
Achtung

Synonyme: Alcianblau 8 GX

12021.01 12021.02
SERA12021.01EA 125 EUR
SERA12021.01 SERA12021.02
Alcianblau 8 GS
Alcianblau 8 GX
Formel: C₅₆H₆₈Cl₄CuN₁₆S₄
Molecular Weight: 1298.86 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00010720
CAS-Nummer: 33864-99-2
EINECS: 251-705-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR