(±)-3-Amino-4,4,4-trifluorbuttersäure 97%

Lieferant: Thermo Scientific
Achtung

L12239.03 L12239.MD
L12239.03EA 114 EUR
L12239.03 L12239.MD
(±)-3-Amino-4,4,4-trifluorbuttersäure 97%
(±)-3-Amino-4,4,4-trifluorbuttersäure

Purity: 97% CAS No.: 584-20-3 Molecular Formula: C4H6F3NO2 Formula Weight: 157.09 Melting Point: ca 184° dec. Beilstein Reference: 1764276

Formel: C₄H₆F₃NO₂
Molecular Weight: 157,09 g/mol
Schmelzpunkt: ∼ 184 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00041413
CAS-Nummer: 584-20-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR