(±)-3-Amino-4,4,4-trifluorbuttersäure
Lieferant: FLUOROCHEM
001477-1G
001477-5G
FLUO001477-1GEA
72.9
EUR
FLUO001477-1G
FLUO001477-5G
(±)-3-Amino-4,4,4-trifluorbuttersäure
(±)-3-Amino-4,4,4-trifluorbuttersäure
Formel:
C₄H₆F₃NO₂ Molecular Weight: 157,09 g/mol |
MDL:
MFCD00041413 CAS-Nummer: 584-20-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...