L-Methioninethylester Hydrochlorid
Lieferant: FLUOROCHEM
Synonyme:
H-Met-OEt·HCl, Ethyl L-methionate hydrochloride
003823-5G
003823-25G
FLUO003823-5GEA
42.5
EUR
FLUO003823-5G
FLUO003823-25G
L-Methioninethylester Hydrochlorid
L-Methioninethylester Hydrochlorid
Formel:
C₇H₁₅NO₂S·HCl Molecular Weight: 213.7 g/mol |
MDL:
MFCD00012508 CAS-Nummer: 2899-36-7 EINECS: 220-787-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...