L(-)-Methioninol 99+%

Lieferant: Thermo Scientific
Achtung

Synonyme: (S)-2-Amino-4-(methylthio)butan-1-ol, L-Methioninol

229230010 229230050
ACRO229230010EA 67.3 EUR
ACRO229230010 ACRO229230050
L(-)-Methioninol 99+%
L(-)-Methioninol
Formel: C₅H₁₃NOS
Molecular Weight: 135,2 g/mol
Schmelzpunkt: 31…33 °C
Flammpunkt: >110 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00004735
CAS-Nummer: 2899-37-8
EINECS: 220-788-1

Bestellen

Spezifikation Testergebnisse

Appearance Transparent white Low melting crystalline solid
Infrared spectrum Conforms
GC ≥99.0 %
Specific optical rotation -11.5° to -13.5° (20°C, 589 nm) (c=1.4, ethanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR