2-Brom-2,2-difluoracetylchlorid
Lieferant: FLUOROCHEM
010428-5G
010428-10G
FLUO010428-5GEA
107
EUR
FLUO010428-5G
FLUO010428-10G
2-Brom-2,2-difluoracetylchlorid
2-Brom-2,2-difluoracetylchlorid
Formel:
C₂BrClF₂O |
MDL:
MFCD00069081 CAS-Nummer: 3832-48-2 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...