Carbohydrazid 97%
Lieferant: Thermo Scientific
|
Achtung
|
403970250
403971000
403975000
ACRO403970250EA
42
EUR
ACRO403970250
ACRO403971000
ACRO403975000
Carbohydrazid 97%
Carbohydrazid
Formel:
CH₆N₄O Molecular Weight: 90,09 g/mol Schmelzpunkt: >153 °C Dichte: 1,02 g/cm³ Lagertemperatur: Kühlschrank |
MDL:
MFCD00007591 CAS-Nummer: 497-18-7 EINECS: 207-837-2 UN: 3077 ADR: 9,III |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 153 to 158 °C |
Titration with KIO3 | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...