tert-Butyl-N-hydroxycarbamat 98+%

Lieferant: Thermo Fisher Scientific

Synonyme: N-Boc-hydroxylamine, N-tert-Butoxycarbonylhydroxylamine, N-Tert-Boc-Hydroxylamine

209870050 209870010 209870250
ACRO209870050EA 92.5 EUR
ACRO209870050 ACRO209870010 ACRO209870250
tert-Butyl-N-hydroxycarbamat 98+%
tert-Butyl-N-hydroxycarbamat
Formel: C₅H₁₁NO₃
Molecular Weight: 133.15 g/mol
Schmelzpunkt: 53…57 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00002107
CAS-Nummer: 36016-38-3
EINECS: 252-836-2

Bestellen

Spezifikation Testergebnisse

Appearance White to light pink Crystalline powder
Infrared spectrum Conforms
Melting point 53 to 57 °C
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR