2-Iodacetylchlorid ≥97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Iodoacetyl chloride

L04428.06 L04428.14
L04428.06EA 108 EUR
L04428.06 L04428.14
2-Iodacetylchlorid ≥97%
2-Iodacetylchlorid

Purity: 97% CAS No.: 38020-81-4 Molecular Formula: ICH2COCl Formula Weight: 204.39 Storage Temperature: 2°C to 8°C UN No.: 3265 MDL No.: MFCD00013718

Formel: C₂H₂ClIO
Molecular Weight: 204,39 g/mol
Siedepunkt: 48…50 °C (12 mmHg)
Dichte: 2,280
MDL: MFCD00013718
CAS-Nummer: 38020-81-4
EINECS: 253-748-7
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR