N-(4-Fluorphenyl)maleamidsäure
Lieferant: FLUOROCHEM
006334-5G
006334-25G
FLUO006334-5GEA
63.9
EUR
FLUO006334-5G
FLUO006334-25G
N-(4-Fluorphenyl)maleamidsäure
N-(4-Fluorphenyl)maleamidsäure
Formel:
C₁₀H₈FNO₃ |
MDL:
MFCD00082643 CAS-Nummer: 60252-79-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...