1-(2-Nitrophenoxy)aceton

Lieferant: BIONET RESEARCH

3J-542S.500 3J-542S.1 3J-542S.5 3J-542S.10
BIOT3J-542S.500EA 0 EUR
BIOT3J-542S.500 BIOT3J-542S.1 BIOT3J-542S.5 BIOT3J-542S.10
1-(2-Nitrophenoxy)aceton
1-(2-Nitrophenoxy)aceton
Formel: C₉H₉NO₄
CAS-Nummer: 5330-66-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR