(p-Toluolsulfonyl)methylisocyanid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1-((Isocyanomethyl)sulphonyl)-4-methylbenzene, Tosylmethyl isocyanide
M-Clarity™ Qualitätsstufe = MQ200
188204-25G
188204-5G
SIAL188204-25GEA
133
EUR
SIAL188204-25G
SIAL188204-5G
(p-Toluolsulfonyl)methylisocyanid, Sigma-Aldrich®
(p-Toluolsulfonyl)methylisocyanid
Formel:
C₉H₉NO₂S Molecular Weight: 195,24 g/mol |
MDL:
MFCD00000005 CAS-Nummer: 36635-61-7 EINECS: 253-140-1 UN: 2811 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...