4-Methoxytritylchlorid (p-Anisylchlordiphenylmethan), Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(Chloro(4-methoxyphenyl)methylene)dibenzene, Mmt-Cl
M-Clarity™ Qualitätsstufe = MQ200
129208-100G
129208-25G
129208-5G
SIAL129208-100GEA
432
EUR
SIAL129208-100G
SIAL129208-25G
SIAL129208-5G
4-Methoxytritylchlorid (p-Anisylchlordiphenylmethan), Sigma-Aldrich®
4-Methoxytritylchlorid (p-Anisylchlordiphenylmethan)
Formel:
C₂₀H₁₇ClO Molecular Weight: 308,8 g/mol |
MDL:
MFCD00000814 CAS-Nummer: 14470-28-1 EINECS: 238-463-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...