(4-Brom-3,5-dimethylphenoxy)acetonitril

Lieferant: COMBI-BLOCKS

HC-3221-5G HC-3221-1G
COBBHC-3221-5GEA 451 EUR
COBBHC-3221-5G COBBHC-3221-1G
(4-Brom-3,5-dimethylphenoxy)acetonitril
(4-Brom-3,5-dimethylphenoxy)acetonitril
Formel: C₁₀H₁₀BrNO
MDL: MFCD09906444
CAS-Nummer: 951918-29-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR