5-Brom-2-benzo[b]thiophencarbonsäure

Lieferant: BIONET RESEARCH

3T-1185.500 3T-1185.1 3T-1185.5
BIOT3T-1185.500EA 0 EUR
BIOT3T-1185.500 BIOT3T-1185.1 BIOT3T-1185.5
5-Brom-2-benzo[b]thiophencarbonsäure
5-Brom-2-benzo[b]thiophencarbonsäure
Formel: C₉H₅BrO₂S
MDL: MFCD01929338
CAS-Nummer: 7312-10-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR