3-Chlor-2-benzo[b]thiophencarbonylchlorid

Lieferant: FLUOROCHEM

Synonyme: 3-Chlorobenzo[b]thiophene-2-carbonyl chloride

019062-5G 019062-25G
FLUO019062-5GEA 155 EUR
FLUO019062-5G FLUO019062-25G
3-Chlor-2-benzo[b]thiophencarbonylchlorid
3-Chlor-2-benzo[b]thiophencarbonylchlorid
Formel: C₉H₄Cl₂OS
Molecular Weight: 231,10 g/mol
MDL: MFCD00053069
CAS-Nummer: 21815-91-8
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR