3-Chlor-2-benzo[b]thiophencarbonylchlorid

Lieferant: Thermo Scientific

Synonyme: 3-Chlorobenzo[b]thiophene-2-carbonyl chloride

BTB00300DA BTB00300EA BTB00300.50
MAYBBTB00300DAEA 0 EUR
MAYBBTB00300DA MAYBBTB00300EA MAYBBTB00300.50
3-Chlor-2-benzo[b]thiophencarbonylchlorid
3-Chlor-2-benzo[b]thiophencarbonylchlorid
Formel: C₉H₄Cl₂OS
Molecular Weight: 231,10 g/mol
MDL: MFCD00053069
CAS-Nummer: 21815-91-8
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR