2-Benzo[b]thiophencarbonylchlorid

Lieferant: Thermo Scientific

Synonyme: Benzo[b]thiophene-2-carbonyl chloride

CC12202DA CC12202EA
MAYBCC12202DAEA 0 EUR
MAYBCC12202DA MAYBCC12202EA
2-Benzo[b]thiophencarbonylchlorid
2-Benzo[b]thiophencarbonylchlorid
Formel: C₉H₅ClOS
Molecular Weight: 196,65 g/mol
MDL: MFCD00051635
CAS-Nummer: 39827-11-7
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR