Ethyl-5-amino-2-benzo[b]thiophencarboxylat

Lieferant: BIONET RESEARCH

8X-0733.500 8X-0733.1 8X-0733.5 8X-0733.10
BIOT8X-0733.500EA 0 EUR
BIOT8X-0733.500 BIOT8X-0733.1 BIOT8X-0733.5 BIOT8X-0733.10
Ethyl-5-amino-2-benzo[b]thiophencarboxylat
Ethyl-5-amino-2-benzo[b]thiophencarboxylat
Formel: C₁₁H₁₁NO₂S
CAS-Nummer: 25785-10-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR