Ethyl-5-nitro-2-benzo[b]thiophencarboxylat

Lieferant: BIONET RESEARCH

12T-0297.500 12T-0297.1 12T-0297.5 12T-0297.10
BIOT12T-0297.500EA 0 EUR
BIOT12T-0297.500 BIOT12T-0297.1 BIOT12T-0297.5 BIOT12T-0297.10
Ethyl-5-nitro-2-benzo[b]thiophencarboxylat
Ethyl-5-nitro-2-benzo[b]thiophencarboxylat
Formel: C₁₁H₉NO₄S
Molecular Weight: 251.258 g/mol
MDL: MFCD03791163
CAS-Nummer: 25785-09-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR