2-Benzo[b]thiophencarbonsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
A12471.03
A12471.06
A12471.14
A12471.03EA
28.6
EUR
A12471.03
A12471.06
A12471.14
2-Benzo[b]thiophencarbonsäure ≥98%
2-Benzo[b]thiophencarbonsäure
Purity: 98%
CAS No.: 6314-28-9
Molecular Formula: C9H6O2S
Formula Weight: 178.22
Melting Point: 236-242°
Beilstein Reference: 124613
Merck Reference: 14,9345
MDL No.: MFCD00051636
Formel:
C₉H₆O₂S Molecular Weight: 178,22 g/mol Schmelzpunkt: 236…242 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051636 CAS-Nummer: 6314-28-9 Merck Index: 14,9345 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...