2-Benzo[b]thiophencarbonsäure

Lieferant: FLUOROCHEM

018198-1G 018198-5G
FLUO018198-1GEA 34.1 EUR
FLUO018198-1G FLUO018198-5G
2-Benzo[b]thiophencarbonsäure
2-Benzo[b]thiophencarbonsäure
Formel: C₉H₆O₂S
MDL: MFCD00051636
CAS-Nummer: 6314-28-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR