3-Benzo[b]thiophencarbonsäure
Lieferant: Thermo Scientific
Synonyme:
1-Benzothiophene-3-carboxylic acid
CC12301.250
CC12301DA
MAYBCC12301.250EA
0
EUR
MAYBCC12301.250
MAYBCC12301DA
3-Benzo[b]thiophencarbonsäure
3-Benzo[b]thiophencarbonsäure
Formel:
C₉H₆O₂S Molecular Weight: 178,22 g/mol |
MDL:
MFCD01846406 CAS-Nummer: 5381-25-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...