5-Amino-2-benzo[b]thiophencarbonsäure

Lieferant: BIONET RESEARCH

9X-0700.1 9X-0700.10 9X-0700.5 9X-0700.500
BIOT9X-0700.1EA 0 EUR
BIOT9X-0700.1 BIOT9X-0700.10 BIOT9X-0700.5 BIOT9X-0700.500
5-Amino-2-benzo[b]thiophencarbonsäure
5-Amino-2-benzo[b]thiophencarbonsäure
Formel: C₉H₇NO₂S
CAS-Nummer: 98589-46-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR