1-(2-Bromphenyl)cyclopropancarbonitril

Lieferant: COMBI-BLOCKS

HC-2961-1G HC-2961-5G
COBBHC-2961-1GEA 303 EUR
COBBHC-2961-1G COBBHC-2961-5G
1-(2-Bromphenyl)cyclopropancarbonitril
1-(2-Bromphenyl)cyclopropancarbonitril
Formel: C₁₀H₈BrN
Molecular Weight: 222,0812 g/mol
MDL: MFCD07374407
CAS-Nummer: 124276-75-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR