1-(3-Bromphenyl)cyclopropancarbonitril

Lieferant: COMBI-BLOCKS

HC-2962-5G HC-2962-1G
COBBHC-2962-5GEA 273 EUR
COBBHC-2962-5G COBBHC-2962-1G
1-(3-Bromphenyl)cyclopropancarbonitril
1-(3-Bromphenyl)cyclopropancarbonitril
Formel: C₁₀H₈BrN
Molecular Weight: 222,0812 g/mol
MDL: MFCD07374414
CAS-Nummer: 124276-83-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR