1-(4-Bromphenyl)cyclopentancarbonitril

Lieferant: COMBI-BLOCKS

HC-3007-1G HC-3007-5G
COBBHC-3007-1GEA 141 EUR
COBBHC-3007-1G COBBHC-3007-5G
1-(4-Bromphenyl)cyclopentancarbonitril
1-(4-Bromphenyl)cyclopentancarbonitril
Formel: C₁₂H₁₂BrN
MDL: MFCD01536834
CAS-Nummer: 143328-19-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR