1-(4-Bromphenyl)cyclopropancarbonitril

Lieferant: COMBI-BLOCKS

CA-4678-1G CA-4678-5G
COBBCA-4678-1GEA 35.8 EUR
COBBCA-4678-1G COBBCA-4678-5G
1-(4-Bromphenyl)cyclopropancarbonitril
1-(4-Bromphenyl)cyclopropancarbonitril
Formel: C₁₀H₈BrN
Molecular Weight: 222.0812 g/mol
MDL: MFCD01314308
CAS-Nummer: 124276-67-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR