1-(4-Chlorphenyl)cyclopropancarbonitril
Lieferant: COMBI-BLOCKS
HC-2915-5G
HC-2915-25G
COBBHC-2915-5GEA
70
EUR
COBBHC-2915-5G
COBBHC-2915-25G
1-(4-Chlorphenyl)cyclopropancarbonitril
1-(4-Chlorphenyl)cyclopropancarbonitril
Formel:
C₁₀H₈ClN Molecular Weight: 177,6302 g/mol |
MDL:
MFCD00019211 CAS-Nummer: 64399-27-5 EINECS: 264-871-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...