4,7-Dichlor-2,8-dimethylchinolin
Lieferant: COMBI-BLOCKS
HC-2145-5G
HC-2145-1G
COBBHC-2145-5GEA
227
EUR
COBBHC-2145-5G
COBBHC-2145-1G
4,7-Dichlor-2,8-dimethylchinolin
4,7-Dichloro-2,8-dimethylquinoline
Formel:
C₁₁H₉Cl₂N Molecular Weight: 226.1000 g/mol |
MDL:
MFCD00272342 CAS-Nummer: 21728-15-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...