5-Brom-4-chlor-3-indolylphosphat p-Toluidinsalz (BCIP (p-Toluidinsalz), X-Phosphat)
Lieferant: Uptima
|
Achtung
|
Synonyme:
BCIP, P-toluidine 5-bromo-4-chloro-1H-indol-3-yl phosphate
UP10904E
UP10904F
UPTIUP10904EEA
0
EUR
UPTIUP10904E
UPTIUP10904F
5-Brom-4-chlor-3-indolylphosphat p-Toluidinsalz (BCIP (p-Toluidinsalz), X-Phosphat)
5-Brom-4-chlor-3-indolylphosphat p-Toluidinsalz (BCIP (p-Toluidinsalz), X-Phosphat)
Formel:
C₁₅H₁₅BrClN₂O₄P Molecular Weight: 433,6214 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00040636 CAS-Nummer: 6578-06-9 EINECS: 229-506-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...