4-Methylumbelliferyl phosphate
Lieferant: Biotium
Synonyme:
Phosphoric Acid 4-Methylumbelliferyl Ester, 4-Methylumbelliferyl phosphoric acid, 4-Methyl-2-oxo-2H-chromen-7-yl dihydrogen phosphate
10009-1
10009
10009-1EA
1130
EUR
10009-1
10009.
4-Methylumbelliferyl phosphate
4-Methylumbelliferyl phosphate
Formel:
C₁₀H₉O₆P Molecular Weight: 256,15 g/mol |
MDL:
MFCD00016969 CAS-Nummer: 3368-04-5 EINECS: 222-137-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...