4-Methylumbelliferyl phosphate

Lieferant: Serva

Synonyme: Phosphoric Acid 4-Methylumbelliferyl Ester, 4-Methylumbelliferyl phosphoric acid, 4-Methyl-2-oxo-2H-chromen-7-yl dihydrogen phosphate

29680.01 29680.02
SERA29680.01EA 0 EUR
SERA29680.01 SERA29680.02
4-Methylumbelliferyl phosphate
4-Methylumbelliferyl phosphate
Formel: C₁₀H₉O₆P
Molecular Weight: 256,15 g/mol
MDL: MFCD00016969
CAS-Nummer: 3368-04-5
EINECS: 222-137-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR