4-Fluor-1-naphthonitril
Lieferant: FLUOROCHEM
Synonyme:
4-Fluoronaphthalene-1-carbonitrile
CFF994.1
004588-1G
004588-5G
004588-25G
CHMPCFF994.1EA
522
EUR
CHMPCFF994.1
FLUO004588-1G
FLUO004588-5G
FLUO004588-25G
4-Fluor-1-naphthonitril
4-Fluor-1-naphthonitril
Formel:
C₁₁H₆FN Molecular Weight: 171.18 g/mol |
MDL:
MFCD00236656 CAS-Nummer: 13916-99-9 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...