2-(3-Brompropoxy)tetrahydropyran

Lieferant: CHEMPUR

Synonyme: 2-(3-Bromopropoxy)tetrahydro-2H-pyran

JFD01859.250 JFD01859.1
CHMPJFD01859.250EA 0 EUR
CHMPJFD01859.250 MAYBJFD01859.1
2-(3-Brompropoxy)tetrahydropyran
2-(3-Brompropoxy)tetrahydropyran
Formel: C₈H₁₅BrO₂
Molecular Weight: 223,1075 g/mol
MDL: MFCD00058593
CAS-Nummer: 33821-94-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR