2-(4-Bromphenoxy)tetrahydropyran

Lieferant: COMBI-BLOCKS

Synonyme: 2-(4-Bromophenoxy)tetrahydro-2H-pyran

OT-0859-5G OT-0859-25G OT-0859-100G
COBBOT-0859-5GEA 0 EUR
COBBOT-0859-5G COBBOT-0859-25G COBBOT-0859-100G
2-(4-Bromphenoxy)tetrahydropyran
2-(4-Bromphenoxy)tetrahydropyran
Formel: C₁₁H₁₃BrO₂
MDL: MFCD00091551
CAS-Nummer: 36603-49-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR