2,3,5-Trichlor-4-formylpyridin
Lieferant: COMBI-BLOCKS
Synonyme:
2,3,5-Trichloroisonicotinaldehyde
SS-3012-5G
SS-3012-1G
COBBSS-3012-5GEA
89.5
EUR
COBBSS-3012-5G
COBBSS-3012-1G
2,3,5-Trichlor-4-formylpyridin
2,3,5-Trichlor-4-formylpyridin
Formel:
C₆H₂Cl₃NO Molecular Weight: 210,4500 g/mol |
MDL:
MFCD06410668 CAS-Nummer: 251997-31-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...