(6-Brom-3-pyridinyl)methanol
Lieferant: COMBI-BLOCKS
Synonyme:
2-Bromo-5-(hydroxymethyl)pyridine, 2-Bromo-5-pyridinemethanol, (6-Bromopyridin-3-yl)methanol
OT-8103-5G
OT-8103-1G
COBBOT-8103-5GEA
101
EUR
COBBOT-8103-5G
COBBOT-8103-1G
(6-Brom-3-pyridinyl)methanol
(6-Brom-3-pyridinyl)methanol
Formel:
C₆H₆BrNO Molecular Weight: 188,02 g/mol |
MDL:
MFCD04974508 CAS-Nummer: 122306-01-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...