(3-Brom-2-pyridinyl)methanol
Lieferant: COMBI-BLOCKS
Synonyme:
3-Bromo-2-(hydroxymethyl)pyridine, (3-Bromopyridin-2-yl)methanol
PY-1423-1G
PY-1423-250M
COBBPY-1423-1GEA
232
EUR
COBBPY-1423-1G
COBBPY-1423-250M
(3-Brom-2-pyridinyl)methanol
(3-Brom-2-pyridinyl)methanol
Formel:
C₆H₆BrNO Molecular Weight: 188,0200 g/mol |
MDL:
MFCD03095348 CAS-Nummer: 52378-64-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...