(5-Brom-2-pyridinyl)methanol

Lieferant: COMBI-BLOCKS

Synonyme: 5-Bromo-2-(hydroxymethyl)pyridine, 5-Bromo-2-hydroxymethylpyridine

PY-7001-25G PY-7001-5G PY-7001-1G
COBBPY-7001-25GEA 352 EUR
COBBPY-7001-25G COBBPY-7001-5G COBBPY-7001-1G
(5-Brom-2-pyridinyl)methanol
(5-Brom-2-pyridinyl)methanol
Formel: C₆H₆BrNO
Molecular Weight: 188.0200 g/mol
MDL: MFCD04035597
CAS-Nummer: 88139-91-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR