(2-Chlor-3-pyridinyl)methanol

Lieferant: BIONET RESEARCH

Synonyme: 2-Chloro-3-pyridinemethanol, (2-chloropyridin-3-yl)methanol, 2-Chloro-3-pyridinylmethanol

2L-375S.500 2L-375S.1 2L-375S.5 2L-375S.10
BIOT2L-375S.500EA 0 EUR
BIOT2L-375S.500 BIOT2L-375S.1 BIOT2L-375S.5 BIOT2L-375S.10
(2-Chlor-3-pyridinyl)methanol
(2-Chlor-3-pyridinyl)methanol
Formel: C₆H₆ClNO
Molecular Weight: 143.57 g/mol
MDL: MFCD01935961
CAS-Nummer: 42330-59-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR