(2-Chlor-4-pyridinyl)methanol

Lieferant: COMBI-BLOCKS

Synonyme: (2-Chloropyridin-4-yl)methanol, 2-Chloro-4-(hydroxymethyl)pyridine, 2-Chloropyridine-4-methanol

PY-1162-5G PY-1162-1G
COBBPY-1162-5GEA 154 EUR
COBBPY-1162-5G COBBPY-1162-1G
(2-Chlor-4-pyridinyl)methanol
(2-Chlor-4-pyridinyl)methanol
Formel: C₆H₆ClNO
Molecular Weight: 143.57 g/mol
MDL: MFCD06858767
CAS-Nummer: 100704-10-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR