(6-Chlor-3-pyridinyl)methanol

Lieferant: BIONET RESEARCH

Synonyme: 6-Chloropyridine-3-methanol, 2-Chloro-5-hydroxymethylpyridine, 6-Chloro-3-(hydroxymethyl)pyridine

5L-348S.500 5L-348S.1 5L-348S.5 5L-348S.10
BIOT5L-348S.500EA 0 EUR
BIOT5L-348S.500 BIOT5L-348S.1 BIOT5L-348S.5 BIOT5L-348S.10
(6-Chlor-3-pyridinyl)methanol
(6-Chlor-3-pyridinyl)methanol
Formel: C₆H₆ClNO
Molecular Weight: 143,57 g/mol
MDL: MFCD01075226
CAS-Nummer: 21543-49-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR