(4-Chlor-2-pyridinyl)methanol
Lieferant: COMBI-BLOCKS
Synonyme:
(4-Chloropyridin-2-yl)methanol, 4-Chloro-2-(hydroxymethyl)pyridine
CA-4132-5G
CA-4132-25G
CA-4132-1G
COBBCA-4132-5GEA
63.4
EUR
COBBCA-4132-5G
COBBCA-4132-25G
COBBCA-4132-1G
(4-Chlor-2-pyridinyl)methanol
(4-Chlor-2-pyridinyl)methanol
Formel:
C₆H₆ClNO Molecular Weight: 143,5709 g/mol |
MDL:
MFCD07437885 CAS-Nummer: 63071-10-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...