Methyl-4-(hydroxymethyl)picolinat 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Methyl 4-(hydroxymethyl)pyridine-2-carboxylate, Methyl 4-(hydroxymethyl)picolinate

H32731.03 H32731.06
H32731.03EA 155 EUR
H32731.03 H32731.06
Methyl-4-(hydroxymethyl)picolinat 95%
Methyl-4-(hydroxymethyl)picolinat

Purity: 95% CAS No.: 317335-15-2 Molecular Formula: C8H9NO3 Formula Weight: 167.16 Melting Point: 74-76° MDL No.: MFCD09834350

Formel: C₈H₉NO₃
Molecular Weight: 167.16 g/mol
Schmelzpunkt: 74…76 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09834350
CAS-Nummer: 317335-15-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR