6-Amino-1,3-dimethyluracil
Lieferant: Thermo Scientific
Synonyme:
6-Amino-1,3-dimethylpyrimidine-2,4(1H,3H)-dione, 4-Amino-1,3-Dimethyluracil
SB01180.1
SB01180.10
SB01180.50
MAYBSB01180.1EA
0
EUR
MAYBSB01180.1
MAYBSB01180.10
MAYBSB01180.50
6-Amino-1,3-dimethyluracil
6-Amino-1,3-dimethyluracil
Formel:
C₆H₉N₃O₂ Molecular Weight: 155,1546 g/mol |
MDL:
MFCD00006552 CAS-Nummer: 6642-31-5 EINECS: 229-662-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...