Riboflavin-5'-phosphat Natriumsalz Dihydrat
Lieferant: Thermo Fisher Scientific
Synonyme:
Sodium (2R,3S,4S)-5-(7,8-dimethyl-2,4-dioxo-3,4-dihydrobenzo[g]pteridin-10(2H)-yl)-2,3,4-trihydroxypentyl hydrogenphosphate dihydrate
A14545.09
A14545.18
A14545.09EA
36.3
EUR
A14545.09
A14545.18
Riboflavin-5'-phosphat Natriumsalz Dihydrat
Riboflavin-5'-phosphat Natriumsalz Dihydrat
CAS No.: 6184-17-4
Molecular Formula: C17H20N4NaO9P.2H2O
Formula Weight: 514.36 (478.34anhy)
Storage Temperature: 2°C to 8°C
Melting Point:
Formel:
C₁₇H₂₀N₄NaO₉P·2H₂O Molecular Weight: 514.36 g/mol (478.34 anhy) Lagertemperatur: Kühlschrank |
MDL:
MFCD00150993 CAS-Nummer: 6184-17-4 EINECS: 204-988-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...